Begerglass, lav modell, Super Duty
Leverandør: DURAN Group
DURAN®, borosilicate glass 3.3, clear, with spout
With easily legible graduation and labelling area. It is recommended to heat the Super Duty glass beakers slowly and evenly to prevent thermal stresses in the glass. For work at very high temperatures or with fast changes in temperature, the standard DURAN® beakers should be used, as these offer excellent resistance to temperature changes. However, mechanical stability is limited with these products in comparison with the Super Duty beakers.
  • Higher mechanical stability compared with standard glass beakers due to reinforced sturdy rim
  • Even distribution of wall thickness
  • Very good chemical resistance, good thermal resistance
With printed retrace code (batch identification); batch certificate available at www.duran-group.com
Logg inn for å se avtalepris og tilgjengelighet
  Husk meg på denne enheten
BESTILL
Utvid alle / Skjul alle
Kapasitet Høyde O.D. Fp Art.nr Antall
1000 ml 145 mm 105 mm 1 214-0207
.
150 ml 80 mm 60 mm 1 214-0199
.
2000 ml 185 mm 132 mm 1 214-0208
.
250 ml 95 mm 70 mm 1 214-0204
.
400 ml 110 mm 80 mm 1 214-0205
.
5000 ml 270 mm 170 mm 1 214-0209
600 ml 125 mm 90 mm 1 214-0206
.
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall