Gloves, Microflex® 93-823
Leverandør: Ansell
Nitrile, non sterile, powder-free, textured fingertips
These disposable gloves made of nitrile provide chemical and liquid protection. The most advanced glove in type I and IV allergy prevention, the Microflex 93-823 has been clinically proven to decrease redness and itchiness associated with contact dermatitis. Engineered without the use of chemical additives (including accelerators) that are known skin irritants, the Microflex 93-823 is safe for those with sensitive skin or prone to allergic reactions. Ideal for use in analytical testing/measurements, assembly and inspection, biotechnologies, cleaning, janitorial and maintenance work, laboratory analysis, pharmaceutical production and light duty assembly.
  • Decreases skin irritation potential in sensitive hands
  • Textured fingertips and a reduced amount of surfactant used to produce the glove results in a secure and confident grip for handling objects
  • Increased sensitivity due to the thin polymer design
  • Ultra-soft materials provide comfort and exceptional fit for long wear periods
EN 374; Cat. III; AQL 1,5
Logg inn for å se avtalepris og tilgjengelighet
  Husk meg på denne enheten
BESTILL
Utvid alle / Skjul alle
Str Farge Lengde Pakket Fp Art.nr Antall
5,5 - 6 Blå 240 mm 100/box 100 112-4321
6,5 - 7 Blå 240 mm 100/box 100 112-4322
7,5 - 8 Blå 240 mm 100/box 100 112-4323
8,5 - 9 Blå 240 mm 100/box 100 112-4324
9,5 - 10 Blå 240 mm 100/box 100 112-4325
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
SPESIFIKASJONER
Modell 93-823
ambidekstral/håndspesifikk Ambidextrous
Engangs/flergangs Disposable
Pudder/pudderfri uten pudder
Overflate Textured fingertips
Type mansjett Perlebesatt
Tykkelse finger 0,10 mm
Palm thickness 0,06 mm
EN norm EN 374
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall