Calibration weight sets, class F1
Leverandør: VWR Collection
Weights made from anti-magnetic, polished stainless steel. Suitable for medium resolution, precision electronic balances. Weights of more than 20 g have adjustment cavities.
  • Wire weights up to 500 mg in cobalt alloy
  • Identification number guarantees traceability for weights from 1 g upwards
  • Laser engraving improves legibility of the markings
  • Available with or without COFRAC certificate
Ordering information: All weight sets listed in the 'set contents' table are available with or without COFRAC certificate - please visit vwr.com or contact your local VWR sales office. Weights are also available individually - please search for product 'Calibration weights, class F1'.
Conform to OILM R111 recommendations.
Logg inn for å se avtalepris og tilgjengelighet
  Husk meg på denne enheten
BESTILL
Utvid alle / Skjul alle
We are currently unable to calculate your contract price for this item. List price is being displayed temporarily. When your order is processed, you will be invoiced at your contract price even though list price is displayed now
F1 weight sets in wooden boxes with certificate
Vektsett Antall vekter Total vekt Fp Art.nr Antall
1 mg - 500 mg 12 1,11 g 1 611-8632
1 mg - 50 g 20 111,11 g 1 611-8633
1 mg - 100 g 21 211,11 g 1 611-8634
1 mg - 200 g 23 611,11 g 1 611-8635
1 mg - 500 g 24 1111,11 g 1 611-8636
1 mg - 1 kg 25 2111,11 g 1 611-8637
1 mg - 2 kg 27 6111,11 g 1 611-8638
1 mg - 5 kg 28 11111,11 g 1 611-8639
1 mg - 10 kg 29 21111,11 g 1 611-8640
1 g - 50 g 8 110 g 1 611-8641
1 g - 100 g 9 210 g 1 611-8642
1 g - 200 g 11 610 g 1 611-8643
1 g - 500 g 12 1110 g 1 611-8644
1 g - 1 kg 13 2110 g 1 611-8645
1 g - 2 kg 15 6110 g 1 611-8646
1 g - 5 kg 16 11110 g 1 611-8647
1 g - 10 kg 17 21110 g 1 611-8648
F1 weight sets in plastic cases with certificate
Vektsett Antall vekter Total vekt Fp Art.nr Antall
1 mg - 500 mg 12 1,11 g 1 611-8461
1 mg - 100 g 21 211,11 g 1 611-8462
1 mg - 200 g 23 611,11 g 1 611-8463
1 mg - 500 g 24 1111,11 g 1 611-8464
1 mg - 2 kg 27 6111,11 g 1 611-8649
1 mg - 5 kg 28 11111,11 g 1 611-8650
1 g - 100 g 9 210 g 1 611-8465
1 g - 200 g 11 610 g 1 611-8466
1 g - 500 g 12 1110 g 1 611-8467
1 g - 2 kg 15 6110 g 1 611-8651
1 g - 5 kg 16 11110 g 1 611-8652
F1 weight sets in wooden boxes without certificate
Vektsett Antall vekter Total vekt Fp Art.nr Antall
1 mg - 500 mg 12 1,11 g 1 611-8602
1 mg - 50 g 20 111,11 g 1 611-8603
1 mg - 100 g 21 211,11 g 1 611-8292
1 mg - 200 g 23 611,11 1 611-8604
1 mg - 500 g 24 1111,11 g 1 611-8605
1 mg - 1 kg 25 2111,11 g 1 611-8606
1 mg - 2 kg 27 6111,11 g 1 611-8607
1 mg - 5 kg 28 11111,11 g 1 611-8608
1 mg - 10 kg 29 21111,11 g 1 611-8609
1 g - 50 g 8 110 g 1 611-8610
1 g - 100 g 9 210 g 1 611-8611
1 g - 200 g 11 610 g 1 611-8612
1 g - 500 g 12 1110 g 1 611-8613
1 g - 1 kg 13 2110 g 1 611-8614
1 g - 2 kg 15 6110 g 1 611-8615
1 g - 5 kg 16 11110 g 1 611-8616
1 g - 10 kg 17 21110 g 1 611-8617
F1 weight sets in plastic cases without certificate
Vektsett Antall vekter Total vekt Fp Art.nr Antall
1 mg - 2 kg 27 6111,11 g 1 611-8618
1 mg - 5 kg 28 11111,11 g 1 611-8619
1 g - 2 kg 15 6110 g 1 611-8620
1 g - 5 kg 16 11110 g 1 611-8621
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall