HPTLC plates, ProteoChrom®
Leverandør: Merck Millipore
ProteoChrom®
For ProteoChrom® HPTLC silica gel 60 F254s plates, an extra thin layer of high performance silica gel is used, providing highly efficient separation characteristics for 1-D analysis of peptides and protein digests. Up to 20 peptides can be resolved, and as little as 1 - 2 ng per band can be seen.
  • Highly reproducible: Optimised separation and staining procedures
  • Convenient, easy to follow, detailed protocols included
  • Highly stable in water, ideal for use with aqueous solvent systems
For ProteoChrom® HPTLC cellulose sheets, an extra thin layer of optimised microcrystalline cellulose is used. Specially developed protocols for development and staining enable a straightforward 2-D analysis in just four hours.
BESTILL
Utvid alle / Skjul alle
Aluminium sheets
Beleggingsmateriale Platestørrelse Layer thickness Porestr Fluorescerende indikator Fp Art.nr Antall
Cellulose 100×100 mm 100 µm 60 Å No 1 1.05651.0001
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall