Capillary columns, VF-17ms
Leverandør: Agilent Technologies
VF-17ms is a 50% phenyl, 50% dimethylpolysiloxane, medium polarity, low bleed column for increased sensitivity, accuracy and instrument uptime.
  • Deactivation technology improves data quality
  • Ideal EPA confirmation column for ultimate confidence
  • Bonded and cross-linked to allow solvent rinsing, reducing replacement costs
  • Improves column stability
BESTILL
Utvid alle / Skjul alle
Column format Phase Tykkhet film Innvendig-Ø×L Fp Art.nr Antall
Capillary VF-17ms 0,15 µm 0,15 mm × 10 m 1 VARICP5882
Capillary VF-17ms 0,20 µm 0,10 mm × 10 m 1 VARICP8977
Capillary VF-17ms 0,15 µm 0,15 mm × 15 m 1 VARICP5883
Capillary VF-17ms 0,25 µm 0,25 mm × 15 m 1 VARICP8979
Capillary VF-17ms 0,50 µm 0,25 mm × 15 m 1 VARICP8980
Capillary VF-17ms 0,15 µm 0,32 mm × 15 m 1 VARICP8986
Capillary VF-17ms 1,00 µm 0,53 mm × 15 m 1 VARICP8996
Capillary VF-17ms 1,50 µm 0,53 mm × 15 m 1 VARICP8998
Capillary VF-17ms 0,15 µm 0,15 mm × 20 m 1 VARICP5884
Capillary VF-17ms 0,05 µm 0,15 mm × 20 m 1 VARICP9009
Capillary VF-17ms 0,15 µm 0,25 mm × 30 m 1 VARICP8981
Capillary VF-17ms 0,25 µm 0,25 mm × 30 m 1 VARICP8982
Capillary VF-17ms 0,25 µm 0,25 mm × 30 m 1 VARICP8982I5
Capillary VF-17ms 0,50 µm 0,25 mm × 30 m 1 VARICP8983
Capillary VF-17ms 0,25 µm 0,32 mm × 30 m 1 VARICP8990
Capillary VF-17ms 0,50 µm 0,32 mm × 30 m 1 VARICP8991
Capillary VF-17ms 1,00 µm 0,53 mm × 30 m 1 VARICP9001
Capillary VF-17ms 1,50 µm 0,53 mm × 30 m 1 VARICP9002
Capillary VF-17ms 0,25 µm 0,25 mm × 60 m 1 VARICP8984
Capillary VF-17ms 0,25 µm 0,32 mm × 60 m 1 VARICP8992
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall