Capillary columns, VF-23ms
Leverandør: Agilent Technologies
VF-23ms features a unique combination of high polarity and low bleed to enable more accurate analysis of very polar analytes.
  • 100% bonded phase permits column rinsing to enhance column lifetime
  • Fast run times improve productivity
  • Operating temperature up to 260°C expands the application range
The enhanced stabilization of the VF-23ms permits splitless injection, column rinsing and temperatures up to 260 °C to be used.
BESTILL
Utvid alle / Skjul alle
Column format Phase Tykkhet film Innvendig-Ø×L Fp Art.nr Antall
Capillary VF-23ms 0,15 µm 0,25 mm × 30 m 1 VARICP8821
Capillary VF-23ms 0,25 µm 0,25 mm × 30 m 1 VARICP8822I5
Capillary VF-23ms 0,25 µm 0,25 mm × 30 m 1 VARICP8822
Capillary VF-23ms 0,25 µm 0,32 mm × 30 m 1 VARICP8827
Capillary VF-23ms 0,50 µm 0,53 mm × 30 m 1 VARICP8831
Capillary VF-23ms 0,25 µm 0,25 mm × 60 m 1 VARICP8824I5
Capillary VF-23ms 0,25 µm 0,25 mm × 60 m 1 VARICP8824
Capillary VF-23ms 0,15 µm 0,32 mm × 60 m 1 VARICP8828
Capillary VF-23ms 0,25 µm 0,32 mm × 60 m 1 VARICP8829
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall