Accessories for Kjeldhal Digestion Systems, Behrotest®
Leverandør: Behr
BESTILL
Utvid alle / Skjul alle
Beskrivelse Type Fp Art.nr Antall
Boiling stones, for Kjeldahl digestion, 100 g - 100 g BEHRB00217914
Process fume evacuator with vacuum pump (40 l/h), BEHROSOG 3, for behrotest InKjel - - BEHRB00217925
Round bottom digestion vessel 250 ml, without NS - - 705-0441
Frame for 4 reaction vessels of 750 ml in the InKjel 475 - - 705-0447
Frame for 4 reaction vessels of 500 ml in the InKjel 450 - - 705-0446
Frame for 6 reaction vessels of 250 ml in the InKjel 625 - - 705-0444
Frame for 12 reaction vessels of 250 ml in the InKjel 1225 - - 705-0445
Fume extraction unit for the InKjel 450 and InKjel 475 - - 705-0450
Fume extraction unit for the InKjel 625 - - 705-0448
Fume extraction unit for the InKjel 1225 - - 705-0449
Round bottom digestion vessel, 500 ml, for InKjel - - 705-0442
Round bottom digestion vessel, 750 ml, for InKjel - 1 705-0443
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall