Serological pipettes, open-ended
Leverandør: Kartell
KART2844EA 2775 NOK
KART2844
Serological pipettes, open-ended
Crystal clear PS.
  • One piece construction without junction to avoid leakage during aspiration and to ensure maximum accuracy and sturdiness
  • Certified non pyrogenic, non haemolytic and non cytotoxic for tissue culture applications
  • Permanent graduation in black foresees an additional volume for extra capacity
  • Sterilised by gamma radiation
Forpakning: Individually packed in a paper/plastic pouch.