HPLC columns, NUCLEOGEL® SUGAR
Leverandør: MACHEREY-NAGEL
NUCLEOGEL®
Polymer based HPLC phases that is sulfonated spherical PS/DVB resins in different ionic forms is ideal for separation of monosaccharides, disaccharides and oligosaccharides, sugar alcohols.
  • Ca2+ form is ideal for separation of mono- and oligosaccharides, sugar alcohols
  • Pb2+ form is ideal for separation of mono- and disaccharides from food and biological samples
  • Na+ form is ideal for separation of oligosaccharides from starch hydrolysates and food
  • Recommended operating temperatures range from 60 to 95 °C with maximum pressure of 70 bar
Separation mechanism includes steric exclusion, ligand exchange and partition effects, ligand exchange being the predominant force, since the hydrated metal ions form strong interactions with the hydroxyl groups of the sample molecules. The intensity of these interactions decreases in the sequence Pb, Ca, Na.
BESTILL
Utvid alle / Skjul alle
Column format Phase Porestr Partikkelstr Innvendig-Ø×L Fp Art.nr Antall
Analytical SUGAR Pb 100 Å 10 µm 7,8×300 mm 1 MANA719530
Analytical SUGAR Ca 100 Å 10 µm 6,5×300 mm 1 MANA719531
Analytical SUGAR Na 100 Å 10 µm 7,8×300 mm 1 MANA719532
Guard columns
Column format Phase Porestr Partikkelstr Innvendig-Ø×L Fp Art.nr Antall
Guard column SUGAR Pb 100 Å 10 µm 4,0×21 mm 2 MANA719534
Guard column SUGAR Ca 100 Å 10 µm 4,0×21 mm 2 MANA719535
Guard column SUGAR Na 100 Å 10 µm 4,0×21 mm 2 MANA719536
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall