Plasma freezers, Revco®
Leverandør: Thermo Scientific
Revco®
These freezers are designed to meet AABB, ANRC and FDA standards for safety and performance in plasma storage. Five models are available with different cabinet sizes to meet a variety of space needs and storage volume requirements.
  • Simplified operation and increased accuracy with the microprocessor control
  • CFC free refrigerant
  • Preset temperature at −30 °C
  • Positive airflow systems
Automatic condensate removal and defrost, with sensor that automatically initiates defrost cycle on demand, eliminating need for a defrost timer, minimizing frost build-up and optimising coil efficiency.
Bestillingsinformasjon: Alarm systems, temperature chart recorders and power monitors are available separately. The additional voltages options are also available on request.
Leveringsinformasjon: Space-saver and upright models come with a 6˝ (15.2 cm) seven-day circular chart temperature recorder (optional free-standing on undercounter model) and extra-long probe for manual alarm testing.
BESTILL
Utvid alle / Skjul alle
Type Fp Art.nr Antall
Plasma freezers, space saver, Revco® UFP1230V 1 REVCUFP1230V
Plasma freezers, undercounter, Revco® UFP430V 1 REVCUFP430V
Plasma freezers, upright, Revco® UFP2330V 1 REVCUFP2330V
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
SPESIFIKASJONER
Modell UFP430V UFP2330V UFP1230V
Kapasitet (L) 133 659 326
Temperatur (°C) −30 °C
Display Digital
Avriming Automatic
Alarm Audible and visual
Hyller/skuffer 2 drawers 7 drawers
Dør(er) 1 solid door
Tilgangsporter 1 (25 mm)
B × D × H inv. (mm) 508×508×508 610×737×1473 508×533×1331
B × D × H utv. (mm) 610×734×864 711×945×2012 610×762×1869
Vekt (kg) 96 193 166
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall