Measuring cylinders, class B
Leverandør: VITLAB
SAN, tall form, class B.
  • Calibrated to contain 'ln'
  • Hexagonal base with bottom studs provides high stability, spout
  • Raised scale and ring marks at the primary scale points
  • Thermal stress up to 60 °C does not cause tolerance limits to be permanently exceeded
Sertifiseringer: DIN 12681 / ISO 6706
BESTILL
Utvid alle / Skjul alle
Kapasitet Inndeling Høyde Toleranse Diam Fp Art.nr Antall
1000 ml 10,0 ml 415 mm 10,0 ± ml 76 mm 1 VITL65291
100 ml 1,0 ml 260 mm 1,00 ± ml 34 mm 1 VITL64991
250 ml 2,0 ml 315 mm 2,00 ± ml 47 mm 1 612-4311
500 ml 5,0 ml 350 mm 5,00 ± ml 61 mm 1 612-4313
50 ml 1,0 ml 199 mm 1,00 ± ml 28 mm 1 VITL64891
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall