Size exclusion chromatography columns, PL aquagel-OH™ preparative
Leverandør: Agilent Technologies
These PL aquagel-OH preparative columns are used in fractionation of wide variety of water-soluble samples based on their size in solution.
  • High loading maximizes sample throughput
  • Up to 10 times scale-up maximizes yield
These columns and associated guard columns enable rapid and convenient scale-up from analytical separations. The columns are packed with the same robust macroporous particles as the analytical column range.
BESTILL
Utvid alle / Skjul alle
Beskrivelse Column format Partikkelstr Innvendig-Ø×L Elimineringsgrense Fp Art.nr Antall
PL aquagel-OH Guard column 10 µm 25×25 mm - 1 VARIPL1249-1120
PL aquagel-OH 30 Preparative 8 µm 25×300 mm 100-30000 g/mol 1 VARIPL1220-6130
PL aquagel-OH 40 Preparative 8 µm 25×300 mm 10000 - 200000 g/mol 1 VARIPL1249-6140
PL aquagel-OH 50 Preparative 8 µm 25×300 mm 50000 - 1000000 g/mol 1 VARIPL1249-6150
PL aquagel-OH 60 Preparative 8 µm 25×300 mm 200000 - >10000000 g/mol 1 VARIPL1249-6160
PL aquagel-OH MIXED Preparative 8 µm 25×300 mm 100 - 10000000 g/mol 1 VARIPL1249-6100
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
TILBEHØR
Utvid alle / Skjul alle
Accessories
Beskrivelse Fp Art.nr Antall
Column connecting nuts, 1/16 in. tube 1 VARIPL1310-0007
Column end plugs, 1/16 inches 1 VARIPL1310-0003
Connecting tubing, 10 cm length, 0,01 in. ID 10 VARIPL1310-0048
Connecting tubing, 50 mm length, 0,25 mm ID 10 VARIPL1310-0047
End fitting for threaded columns, 7,5 mm ID 1 VARIPL1310-0004
Frit kit, 10 μm, for threaded columns, 7,5 mm ID 5 VARIPL1310-0036
Frit kit, 2 μm, for threaded columns, 7,5 mm ID 5 VARIPL1310-0002
Frit kit, 5 μm, for narrow bore MiniMIX columns, 4,6 mm ID 1 VARIPL1310-0042
Frit kit, 5 μm, for threaded columns, 7,5 mm ID 5 VARIPL1310-0012
Frit removal tool for threaded columns only 1 VARIPL1310-0001
LDV intercolumn stainless steel connector 1 VARIPL1310-0005
MesoPore column repair gel 1 VARIPL1413-0325
OligoPore column repair gel 1 VARIPL1413-0320
PLgel column repair gel, 10 μm 1 VARIPL1410-0101
PLgel column repair gel, 20 μm 1 VARIPL1410-0201
PLgel column repair gel, 3 μm 1 VARIPL1410-0301
PLgel column repair gel, 5 μm 1 VARIPL1410-0501
PLgel Olexis column repair gel 1 VARIPL1410-0200
PolyPore column repair gel 1 VARIPL1413-0500
ResiPore column repair gel 1 VARIPL1413-0300
Tubing ferrules, 1/16 in. tube 5 VARIPL1310-0008
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall