Wash bottles, narrow or wide neck
Leverandør: VITLAB
PP, translucent, screw cap and spray tube made from PP.
  • Very good chemical resistance
  • Very good temperature resistance
Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube.
BESTILL
Utvid alle / Skjul alle
Kapasitet Høyde Gjenge Ytre diam Fp Art.nr Antall
250 ml 146 mm GL 45 58 mm 1 VITL93793
500 ml 166 mm GL 45 76 mm 1 VITL93993
1000 ml 226 mm GL 63 91 mm 1 VITL94193
1000 ml 215 mm GL 32 92 mm 1 VITL95193
250 ml 135 mm GL 25 58 mm 1 VITL94993
500 ml 180 mm GL 25 74 mm 1 VITL95093
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall