Flow meter tubes for air
Leverandør: GLASS PRECISION ENGINEERING
Borosilicate glass tubes with linear or direct reading scale giving air or water flow measurements. Clear indelibly fired-in markings and yellow backing give ease of reading. The tubes are made of heat-resisting glass with heavy walls.
  • Repeatability ±0,25 % of full scale reading
  • Rib guided flow tube for maximum stability and accuracy
  • Polyethylene connector
Leveringsinformasjon: Supplied with push-on plastic tube connector.
BESTILL
Utvid alle / Skjul alle
Beskrivelse Strømningshastighet Nøyaktighet Lengde Fp Art.nr Antall
Meterate tube and glass float 0,003 - 0,03 l/min ±0,5 % 150 mm 1 GLPR10301
Meterate tube and ruby float 0,006 - 0,06 l/min ±0,5 % 150 mm 1 GLPR10302
Meterate tube and glass float 0,009 - 0,09 l/min ±0,5 % 150 mm 1 GLPR10303
Meterate tube and stainless steel float 0,01 - 0,1 l/min ±0,5 % 150 mm 1 GLPR10304
Meterate tube and ruby float 0,01 - 0,1 l/min ±0,5 % 150 mm 1 GLPR10305
Meterate tube and stainless steel float 0,03 - 0,3 l/min ±0,5 % 150 mm 1 GLPR10306
Meterate tube and glass float 0,04 - 0,4 l/min ±0,5 % 150 mm 1 GLPR10307
Meterate tube and ruby float 0,05 - 0,5 l/min ±0,5 % 150 mm 1 GLPR10308
Meterate tube and stainless steel float 0,08 - 0,8 l/min ±0,5 % 150 mm 1 GLPR10309
Meterate tube and glass float 0,09 - 0,9 l/min ±0,5 % 150 mm 1 GLPR10310
Meterate tube and ruby float 0,1 - 1 l/min ±0,5 % 150 mm 1 GLPR10311
Meterate tube and stainless steel float 0,2 - 2 l/min ±0,5 % 150 mm 1 GLPR10312
Meterate tube and glass float 0,2 - 2 l/min ±0,5 % 150 mm 1 GLPR10313
Meterate tube and ruby float 0,3 - 3,1 l/min ±0,5 % 150 mm 1 GLPR10314
Meterate tube and glass float 0,4 - 4 l/min ±0,5 % 150 mm 1 GLPR10315
Meterate tube and stainless steel float 0,5 - 5 l/min ±0,5 % 150 mm 1 GLPR10316
Meterate tube and ruby float 0,5 - 5 l/min ±0,5 % 150 mm 1 GLPR10317
Meterate tube and glass float 0,6 - 6 l/min ±0,5 % 150 mm 1 GLPR10318
Meterate tube and ruby float 0,7 - 7 l/min ±0,5 % 150 mm 1 GLPR10319
Meterate tube and stainless steel float 0,8 - 8 l/min ±0,5 % 150 mm 1 GLPR10320
Meterate tube and glass float 0,9 - 9 l/min ±0,5 % 150 mm 1 GLPR10321
Meterate tube and stainless steel float 0,1 - 11 l/min ±0,5 % 150 mm 1 GLPR10322
Meterate tube and ruby float 0,1 - 11 l/min ±0,5 % 150 mm 1 GLPR10323
Meterate tube and stainless steel float 0,2 - 17 l/min ±0,5 % 150 mm 1 GLPR10324
Meterate tube and glass float 0,2 - 20 l/min ±0,5 % 150 mm 1 GLPR10325
Meterate tube and ruby float 0,2 - 23 l/min ±0,5 % 150 mm 1 GLPR10326
Meterate tube and stainless steel float 0,4 - 39 l/min ±0,5 % 150 mm 1 GLPR10327
Meterate tube and hi-denl float 0,3 - 60 l/min ±0,5 % 150 mm 1 GLPR10328
Meterate tube and glass float 0,003 - 0,03 l/min ±0,5 % 150 mm 1 GLPR10401
Meterate tube and ruby float 0,006 - 0,06 l/min ±0,5 % 150 mm 1 GLPR10402
Meterate tube and glass float 0,009 - 0,09 l/min ±0,5 % 150 mm 1 GLPR10403
Meterate tube and stainless steel float 0,01 - 0,1 l/min ±0,5 % 150 mm 1 GLPR10404
Meterate tube and ruby float 0,01 - 0,1 l/min ±0,5 % 150 mm 1 GLPR10405
Meterate tube and stainless steel float 0,03 - 0,3 l/min ±0,5 % 150 mm 1 GLPR10406
Meterate tube and glass float 0,04 - 0,4 l/min ±0,5 % 150 mm 1 GLPR10407
Meterate tube and ruby float 0,05 - 0,5 l/min ±0,5 % 150 mm 1 GLPR10408
Meterate tube and stainless steel float 0,08 - 0,8 l/min ±0,5 % 150 mm 1 GLPR10409
Meterate tube and glass float 0,09 - 0,9 l/min ±0,5 % 150 mm 1 GLPR10410
Meterate tube and ruby float 0,1 - 1 l/min ±0,5 % 150 mm 1 GLPR10411
Meterate tube and stainless steel float 0,2 - 2 l/min ±0,5 % 150 mm 1 GLPR10412
Meterate tube and glass float 0,2 - 2 l/min ±0,5 % 150 mm 1 GLPR10413
Meterate tube and ruby float 0,3 - 3,1 l/min ±0,5 % 150 mm 1 GLPR10414
Meterate tube and glass float 0,4 - 4 l/min ±0,5 % 150 mm 1 GLPR10415
Meterate tube and stainless steel float 0,5 - 5 l/min ±0,5 % 150 mm 1 GLPR10416
Meterate tube and ruby float 0,5 - 5 l/min ±0,5 % 150 mm 1 GLPR10417
Meterate tube and glass float 0,6 - 6 l/min ±0,5 % 150 mm 1 GLPR10418
Meterate tube and ruby float 0,7 - 7 l/min ±0,5 % 150 mm 1 GLPR10419
Meterate tube and stainless steel float 0,8 - 8 l/min ±0,5 % 150 mm 1 GLPR10420
Meterate tube and glass float 0,9 - 9 l/min ±0,5 % 150 mm 1 GLPR10421
Meterate tube and stainless steel float 1 - 11 l/min ±0,5 % 150 mm 1 GLPR10422
Meterate tube and ruby float 1 - 11 l/min ±0,5 % 150 mm 1 GLPR10423
Meterate tube and stainless steel float 2 - 17 l/min ±0,5 % 150 mm 1 GLPR10424
Meterate tube and glass float 2 - 20 l/min ±0,5 % 150 mm 1 GLPR10425
Meterate tube and ruby float 2 - 23 l/min ±0,5 % 150 mm 1 GLPR10426
Meterate tube and stainless steel float 4 - 39 l/min ±0,5 % 150 mm 1 GLPR10427
Meterate tube and hi-den float 0,3 - 60 l/min ±0,5 % 150 mm 1 GLPR10428
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall