Vårt Oslo-kontor flytter den 25. mars 2019 til Brynsalleen 4, 0667 Oslo
Chiral HPLC columns, NUCLEOSIL® CHIRAL
Leverandør: MACHEREY-NAGEL
NUCLEOSIL®
Silica based phases for HPLC enantiomer separation in which CHIRAL-1 seperation is based on ligand exchange and CHIRAL-2 or CHIRAL-3 on charge-transfer and further interactions as well as on steric effects.
CHIRAL-1 based on an L-proline copper complex covalently bonded to spherical silica particles. The formation of ternary mixed ligand complexes via Cu(II) ions with the analytes and differences in the stability of the diastereomeric complexes cause the chromatographic separation. The eluent in the column is a 0,5 mM copper sulfate solution.
The column has recommended applications on enantiomers with two polar functional groups with a suitable distance such as α-amino acids, α-hydroxycarboxylic acids and N-alkyl-α-amino acids.
CHIRAL-2 is a so-called "brush type" phase. The chiral selector N-(3,5-dinitrobenzoyl)-D-phenylglycine is covalently bonded to spherical silica.
CHIRAL-3 features the L-conformation of the chiral selector.
The interaction modes involve charge-transfers, dipole-dipole interactions, hydrogen bonds and steric effects. The eluent in the column is n-heptane/2-propanol/TFAA (100:0.05:0.05, v/v/v).
Separations of enantiomers and diastereomers, the control of the optical purity of plant protectives and pharmaceuticals as well as product controls in chiral organic syntheses are recommended applications of the both columns.
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).
BESTILL
Utvid alle / Skjul alle
Analytical columns
Column format Phase Porestr Partikkelstr Innvendig-Ø×L Fp Art.nr Antall
Analytical NUCLEOSIL® CHIRAL-1 120 Å 5 µm 4,0×250 mm 1 MANA720081.40
Analytical NUCLEOSIL® CHIRAL-2 100 Å 5 µm 4,0×250 mm 1 MANA720088.40
Analytical NUCLEOSIL® CHIRAL-3 100 Å 5 µm 4,0×250 mm 1 MANA720350.40
Guard columns
Column format Phase Porestr Partikkelstr Innvendig-Ø×L Fp Art.nr Antall
Guard column NUCLEOSIL® CHIRAL-1 120 Å 5 µm 3,0×4 mm 3 MANA721188.30
Guard column NUCLEOSIL® CHIRAL-2/3 100 Å 5 µm 3,0×4 mm 3 MANA721190.30
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
SPESIFIKASJONER
Beskrivelse NUCLEOSIL® CHIRAL-2 NUCLEOSIL® CHIRAL-1 NUCLEOSIL® CHIRAL-3 NUCLEOSIL® CHIRAL-2/3 NUCLEOSIL® CHIRAL-1
USP kode L36 L32 L36 L32
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall