Vårt Oslo-kontor flytter den 25. mars 2019 til Brynsalleen 4, 0667 Oslo
Chiral HPLC columns, NUCLEODEX
Leverandør: MACHEREY-NAGEL
Cyclodextrin modified silica phases for HPLC enantiomer separation. These phases are especially suited for the control of optical purity and for the analysis of positional and cis-trans isomers.
NUCLEODEX β-OH is a β-cyclodextrin bonded to porous silica particles. Its separation based on hydrogen bonds and dipole interactions between functional groups of the analyte and hydroxyl groups of the cyclodextrin. Examples for successful enantiomer separations: chlorthalidone and other compounds, which require free hydroxyl groups for enantioselective interactions. Eluent in column CH₃OH - 0,1% TEAA, pH 4.
NUCLEODEX α-PM is a permethylated α-cyclodextrin phase. For all permethylated phases the ability to form hydrogen bonds is reduced, and the hydrophobicity of the phase is increased compared to β‑OH that results in shorter retention times. Examples for successful enantiomer separations: mecoprop and dichlorprop as free carboxylic acids, trans-stilbene oxide and styrene oxide. Eluent in column CH₃OH - 50 mM phosphate, pH 3.
NUCLEODEX β-PM is a permethylated β-cyclodextrin phase ideal for successful enantiomer separations such as mephobarbital (prominal), pesticide derivatives mecoprop-methyl and fenoxaprop-ethyl. Eluent in column CH3OH - 0,1% TEAA, pH 4.
NUCLEODEX γ-PM is a permethylated γ-cyclodextrin phase ideal for successful enantiomer separations such as steroids or other larger molecules. Eluent in column CH₃OH - 0,1% TEAA, pH 4.
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).
BESTILL
Utvid alle / Skjul alle
Analytical columns
Column format Phase Porestr Partikkelstr Innvendig-Ø×L Fp Art.nr Antall
Analytical NUCLEODEX β-OH 100 Å 5 µm 4,0×200 mm 1 MANA720124.40
Analytical NUCLEODEX α-PM 100 Å 5 µm 4,0×200 mm 1 MANA720127.40
Analytical NUCLEODEX β-PM 100 Å 5 µm 4,0×200 mm 1 MANA720125.40
Analytical NUCLEODEX γ-PM 100 Å 5 µm 4,0×200 mm 1 MANA720752.40
Guard columns
Column format Phase Porestr Partikkelstr Innvendig-Ø×L Fp Art.nr Antall
Guard column NUCLEODEX ß-OH 100 Å 5 µm 3,0×4 mm 1 MANA721171.30
Guard column NUCLEODEX α-PM 100 Å 5 µm 3,0×4 mm 3 MANA721469.30
Guard column NUCLEODEX β-PM 100 Å 5 µm 3,0×4 mm 3 MANA721176.30
Guard column NUCLEODEX γ-PM 100 Å 5 µm 3,0×4 mm 3 MANA721178.30
Screening kit
Beskrivelse Fp Art.nr Antall
NUCLEODEX screening kit consists of one CC 30/4 each with NUCLEODEX β-OH, α-PM, β-PM and γ-PM and a CC column holder 30 mm 1 KIT MANA721920
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall