TLC plates, aluminium oxide, Alox
Leverandør: MACHEREY-NAGEL
TLC plates and sheets are used for the separations of alkaloids, steroids, aliphatic and aromatic compounds.
  • Alternative selectivity to silica 60
Aluminium oxide: Specific surface (BET) ~ 200 m²/g
Indicator: Manganese activated zinc silicate
BESTILL
Utvid alle / Skjul alle
Glass plates
Beskrivelse Beleggingsmateriale Platestørrelse Layer thickness Porestr Fluorescerende indikator Binder Fp Art.nr Antall
Alox-25 UV254 Aluminium oxide (Alox) 50×200 mm 250 µm 60 Å F254 Inert organic 100 552-0169
Alox-25 UV254 Aluminium oxide (Alox) 200×200 mm 250 µm 60 Å F254 Inert organic 25 552-0170
Alox-100 UV254 Aluminium oxide (Alox) 200×200 mm 1000 µm 60 Å F254 Inert organic 15 552-0171
POLYGRAM® polyester sheets
Beskrivelse Beleggingsmateriale Platestørrelse Layer thickness Porestr Fluorescerende indikator Binder Fp Art.nr Antall
Alox N/UV254 Aluminium oxide (Alox) 40×80 mm 200 µm 60 Å F254 Inert organic 50 293-5210
Alox N/UV254 Aluminium oxide (Alox) 50×200 mm 200 µm 60 Å F254 Inert organic 50 552-0149
Alox N/UV254 Aluminium oxide (Alox) 200×200 mm 200 µm 60 Å F254 Inert organic 25 552-0150
ALUGRAM® aluminium sheets
Beskrivelse Beleggingsmateriale Platestørrelse Layer thickness Porestr Fluorescerende indikator Binder Fp Art.nr Antall
Alox N/UV254 Aluminium oxide (Alox) 50×200 mm 200 µm 60 Å F254 Inert organic 50 552-0292
Alox N/UV254 Aluminium oxide (Alox) 200×200 mm 200 µm 60 Å F254 Inert organic 25 552-0291
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall