TLC plates, special layers, CHIRALPLATE
Leverandør: MACHEREY-NAGEL
These glass plates with a special layer are used for TLC enantiomer separations (e.g. amino acids, thiazolidine derivatives, dipeptides, lactones, α-hydroxycarboxylic acids).
  • Chiral separation based on ligand exchange
Base material: Reversed phase nano silica
Impregnation: Cu(II) ions and as chiral selector a proline derivative
BESTILL
Utvid alle / Skjul alle
Beskrivelse Beleggingsmateriale Platestørrelse Layer thickness Fluorescerende indikator Fp Art.nr Antall
CHIRALPLATE Silica gel 50×200 mm 250 µm F254 50 552-0239
CHIRALPLATE Silica gel 100×100 mm 250 µm F254 25 552-0241
CHIRALPLATE Silica gel 100×200 mm 250 µm F254 4 552-0238
CHIRALPLATE Silica gel 100×200 mm 250 µm F254 25 552-0237
CHIRALPLATE Silica gel 200×200 mm 250 µm F254 25 552-0240
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall