Calcium disilicide Ca ∼30% may contain up to 5% Fe, tech.
Leverandør: Thermo Scientific
|
Danger
|
14676.09EA
565
NOK
14676.09
14676.36
14676.22
Calcium disilicide Ca ∼30% may contain up to 5% Fe, tech.
Calcium disilicide
Formel:
CaSi₂ MW: 96,25 g/mol Smeltepunkt: ∼ 1000 °C Tetthet: 2,5 Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00015977 CAS-nummer: 12013-56-8 EINECS: 234-588-7 FN: 1405 ADR: 4.3,II |