(±)-2-Metyl-2,4-pentandiol NF
|
Warning
|
N869-500MLEA
915
NOK
N869-500ML
(±)-2-Metyl-2,4-pentandiol NF
(±)-2-Metyl-2,4-pentandiol
Formel:
C₆H₁₄O₂ MW: 118,18 g/mol Kokepunkt: 198 °C (1013 hPa) Smeltepunkt: –40 °C Tetthet: 0,923 g/cm³ (20 °C) Flash punkt: 93 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00004547 CAS-nummer: 107-41-5 EINECS: 203-489-0 Merck Indeks: 13,04730 |
Spesifikasjon Testresultater
Acidity Test | Pass |
Appearance | Clear, colorless liquid |
Refractive Index | 1.424 – 1.43 |
Specific Gravity | 0.917 – 0.923 |
Water | ≤0.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...