Pipetter, Enkanals makro, Calibra® digital 832
Leverandør: Socorex
Calibra® digital
613-4878EA 0 NOK
613-4878
Pipetter, Enkanals makro, Calibra® digital 832
Single channel macropipettes with winding-free, instant volume adjustment.
  • Long term performance stability
  • Shock, heat, chemicals and UV-light resistance.
  • Autoclavable at 121 °C (fully assembled)