Hexafluorophosphoric acid ∼60% (w/w) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16940-81-1
:
Formula:HF₆P
MDL Number:MFCD00011350
:145.97 g/mol
Density:1.65
Storage Temperature:Ambient
EINECS:241-006-5
UN:1782
ADR:8,II
:GHS06
:GHS05
Related Information