Norway
Order Entry
Serum collection tubes, VACUETTE®
Serum collection tubes, VACUETTE®
Supplier:  Greiner Bio-One
Certificates
Serum collection tubes, VACUETTE®
With PREMIUM (Safety twist cap)
Product Information
  • Catalog #
    Supplier #
    Material
    Additive
    Length
    Ø
    Cap colour
    Availability
    Price
  • Blood Collection tube, Vacuette®, 3,5 ml
    Blood Collection tube, Vacuette®, 3,5 ml
    GREI454214_1200
    454214_1200
    PET
    Z serum clot activator
    75 mm
    13 mm
    Gold
    Log in for availability
    1 * 1.200 Items
    1 * 1.200 Items - kr 2 925,00
    kr 2 925,00
    1 * 1.200 Items
       kr 2 925,00
     : 454214_1200
     : GREI454214_1200
    Specifications
    Environmentally Preferable
With non-ridged (pull cap)
  •  
    Material
    Additive
    Length
    Ø
    Cap colour
  • Blood Collection tube, Vacuette®, 3,5 ml
    Blood Collection tube, Vacuette®, 3,5 ml
    GREI454067_1200
    454067_1200
    PET
    Z serum sep clot activator
    75 mm
    13 mm
    Red
    1 * 1.200 Items
    1 * 1.200 Items - kr 2 925,00
    kr 2 925,00
    1 * 1.200 Items
       kr 2 925,00
     : 454067_1200
     : GREI454067_1200
    Specifications
    Environmentally Preferable
  • Blood Collection tube, Vacuette®, 4 ml
    Blood Collection tube, Vacuette®, 4 ml
    GREI454204_1200
    454204_1200
    PET
    Z serum clot activator
    75 mm
    13 mm
    Red
    1 * 1.200 Items
    1 * 1.200 Items - kr 2 200,00
    kr 2 200,00
    1 * 1.200 Items
       kr 2 200,00
     : 454204_1200
     : GREI454204_1200
    Specifications
    Environmentally Preferable
  • Blood Collection tube, Vacuette®, 6 ml
    Blood Collection tube, Vacuette®, 6 ml
    GREI456089_1200
    456089_1200
    PET
    Z serum clot activator
    100 mm
    13 mm
    Red
    1 * 1.200 Items
    1 * 1.200 Items - kr 2 350,00
    kr 2 350,00
    1 * 1.200 Items
       kr 2 350,00
     : 456089_1200
     : GREI456089_1200
    Specifications
    Environmentally Preferable
  • Blood Collection tube, Vacuette®, 3,5 ml
    Blood Collection tube, Vacuette®, 3,5 ml
    GREI454228_1200
    454228_1200
    PET
    Z serum clot activator
    75 mm
    13 mm
    Gold
    1 * 1.200 Items
    1 * 1.200 Items - kr 2 925,00
    kr 2 925,00
    1 * 1.200 Items
       kr 2 925,00
     : 454228_1200
     : GREI454228_1200
    Specifications
    Environmentally Preferable