Anion exchange chromatography columns, HiTrap™ Q FF
Supplier: Cytiva
Certificates
About this item
HiTrap Q FF is prepacked with Q Sepharose Fast Flow, and is a strong anion exchanger for small-scale protein purifications as well as screening of binding and elutions conditions.
- Convenient and affordable for fast, easy ion exchange separations either alone or connected in series
- The industry standard for ion exchange chromatography
- High flow rates and good scale-up potential
- Use a weak ion exchanger if the selectivity of a strong ion exchanger is insufficient
- Predictable scale-up
Composed of crosslinked 6% agarose beads with quaternary amine strong anion exchange groups, this resin has high chemical stability, allowing well proven cleaning-in-place (CIP) and sanitization protocols. The resin’s 90 µm bead size yields rapid flow rates, making it simple to scale up the purification.
Specifications
- Catalog No:
- 17-5053-01
- 17-5156-01
- Max. pressure:
- 5 bar
- 5 bar
- pH range:
- 2 - 12
- 2 - 12
Specifications
Related Information
Product Information
- Catalog #Supplier #Column hardwareParticle SizeBed dimensionsCapacityFlow rateAvailabilityPrice
- HisTrap™ Q FFHisTrap™ Q FF17-5053-0117-5053-01PP90 µm7×25 mm1 ml<4 ml/minLog in for availability1 * 5 Itemskr 1 925,00Supplier Article Number: 17-5053-01Local Article Number: GEHE17-5053-01Specifications:
More
Specifications
Environmentally PreferableMax. pressure5 barpH range2 - 12 - HisTrap™ Q FFHisTrap™ Q FF17-5156-0117-5156-01PP90 µm16×25 mm5 ml<20 ml/minLog in for availability1 * 5 Itemskr 4 175,00Supplier Article Number: 17-5156-01Local Article Number: GEHE17-5156-01Specifications:
More
Specifications
Environmentally PreferableMax. pressure5 barpH range2 - 12