Select-a-dye mono-reactive dye packs
Leverandør: Cytiva
PA15123EA 4575 NOK
PA15123
Select-a-dye mono-reactive dye packs
Systemer for proteinkobling og -merking
Select-a-dye mono-reactive dye packs enables a variety of labelling strategies using NHS ester, maleimide, or hydrazide to be investigated. Example protocols included with each dye pack.
Leveringsinformasjon: Contains 0,5 mg of each labelling chemistry per pack.