IBC roll top hardcover, PIG®
Leverandør: New Pig
PIG®
NEWPPAK902-BWG-AWOEA 43975 NOK
NEWPPAK902-BWG-AWO
IBC roll top hardcover, PIG®
Materialer for håndtering av søl
This roll top provides protected storage and liquid containment for IBC units. The LDPE construction with UV inhibitors resists UV rays, rust, corrosion and most chemicals.
  • Weather-resistant
  • Two hinged doors and one sliding door
  • Two-way forklift entry for easy movement
  • Durable rolling cover and swing-out doors on both sides provide easy access
  • 14% post-industrial recycled polyethylene
  • Roll top can be secured with a padlock (sold separately)
Low-density polyethylene (LDPE) construction with UV inhibitors resists UV rays, rust, corrosion and most chemicals.
Weight: 242,9 kg