Tris-glycine buffer, 10X concentrate, Proteomics Grade
M114-4LEA 0 NOK
VWRCM114-4L
Tris-glycine buffer, 10X concentrate, Proteomics Grade
A 1X working solution contains 0,025 M Tris base, 0,192 M glycine. 1X buffer is used as a transfer buffer for Western blotting procedures.