Welding spectacles, Eagle
Leverandør: 3M
111-1070EA 350 NOK
MMMAEAG3.0S
Welding spectacles, Eagle
Available with either plastic lenses (CR39) or PC lenses. The PC lenses eliminate UV light, reduce infra-red rays and ensure optimum protection for the eyes by controlling visible light. The spectacles can be used anywhere where UV, IR or visible light poses a risk. Available in shades ranging from 1,7 (welder's assistant) to 7,0 (oxyacetylene cutting).
  • Integrated side shields with air vents
  • Adjustable earpiece length (four levels)
  • Frame angle can be adjusted to three different positions
  • Rotatable earpiece ends for a perfect fit
Sertifiseringer: EN 166