HPLC columns, Hypersil GOLD™ PFP
Efficient separation of difficult mixtures containing closely related halogenated and polar compounds. The introduction of a fluorine group into the stationary phase causes significant changes in solute-stationary phase interaction and increases the selectivity and extra retention.
- Increased retention and selectivity for positional isomers of halogenated compounds
- Excellent selective analysis of non halogenated compounds - in particular polar compounds containing hydroxyl, carboxyl, nitro, or other polar groups
Hypersil GOLD™ PFP columns
- PhaseColumn FormatPore sizeParticle SizeØ int.Length
- 554-431125402-052130PFP175 Å1,9 µm2,10 mm50 mm1 * 1 Itemskr 10 561,25
Specifications
Environmentally Preferable - 555-560725402-102130PFP175 Å1,9 µm2,10 mm100 mm1 * 1 Itemskr 10 646,25
Specifications
Environmentally Preferable - 555-561025402-152130PFP175 Å1,9 µm2,10 mm150 mm1 * 1 Itemskr 11 602,50
Specifications
Environmentally Preferable - 555-557525403-052130PFP175 Å3 µm2,10 mm50 mm1 * 1 Itemskr 11 625,00
Specifications
Environmentally Preferable - 555-558325403-102130PFP175 Å3 µm2,10 mm100 mm1 * 1 Itemskr 12 650,00
Specifications
Environmentally Preferable - 555-559125403-152130PFP175 Å3 µm2,10 mm150 mm1 * 1 Itemskr 13 375,00
Specifications
Environmentally Preferable - 555-558625403-104630PFP175 Å3 µm4,60 mm100 mm1 * 1 Itemskr 9 817,50
Specifications
Environmentally Preferable - 554-431225403-154630PFP175 Å3 µm4,60 mm150 mm1 * 1 Itemskr 10 327,50
Specifications
Environmentally Preferable - 555-562825405-052130PFP175 Å5 µm2,10 mm50 mm1 * 1 Itemskr 10 575,00
Specifications
Environmentally Preferable - 555-549825405-152130PFP175 Å5 µm2,10 mm150 mm1 * 1 Itemskr 12 175,00
Specifications
Environmentally Preferable - 555-550125405-154630PFP175 Å5 µm4,60 mm150 mm1 * 1 Itemskr 9 413,75
Specifications
Environmentally Preferable - 554-431325405-254630PFP175 Å5 µm4,60 mm250 mm1 * 1 Itemskr 9 923,75
Specifications
Environmentally Preferable
Drop-in guard cartridges
- PhaseColumn FormatPore sizeParticle SizeØ int.Length
- 555-561425403-011001PFPGuard cartridge175 Å3 µm1,00 mm10 mm1 * 4 Itemskr 7 175,00
Specifications
Environmentally Preferable - 555-561525403-012101PFPGuard cartridge175 Å3 µm2,10 mm10 mm1 * 4 Itemskr 6 725,00
Specifications
Environmentally Preferable - 555-561725403-013001PFPGuard cartridge175 Å3 µm3,00 mm10 mm1 * 4 Itemskr 7 175,00
Specifications
Environmentally Preferable - 555-561825403-014001PFPGuard cartridge175 Å3 µm4,00/4,60 mm10 mm1 * 4 Itemskr 7 175,00
Specifications
Environmentally Preferable - 555-565325405-012101PFPGuard cartridge175 Å5 µm2,10 mm10 mm1 * 4 Itemskr 6 425,00
Specifications
Environmentally Preferable - 555-565525405-014001PFPGuard cartridge175 Å5 µm4,00/4,60 mm10 mm1 * 4 Itemskr 6 125,00
Specifications
Environmentally Preferable