Carbamazepine
Leverandør: LGC Standards PROMOCHEM
PROOICRS0239EA
0
NOK
PROOICRS0239
Carbamazepine
Carbamazepine
Formel:
C₁₅H₁₂N₂O MW: 236,27 g/mol |
MDL nummer:
MFCD00005073 CAS-nummer: 298-46-4 EINECS: 206-062-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...