Flow indicator
Leverandør: SciLabware
AZLOFWY053EA 970 NOK
AZLOFWY053
Flow indicator
Maximum operating temperature: 60 °C
  • Indicates flow of liquid or gas
  • Transparent robust PMP moulding with coloured paddle wheel
  • Operates at low flow rates - the impeller starts to turn at approximately 150 ml/minute
NB: Not for use with some organic solvents, strong acids or strong bases. Please check compatibility of flow materials with PMP at relevant temperature(s).