Safety ankle boots for ladies, lace-up, Temptation® City Top
Leverandør: Honeywell Safety Products
114-0724EA 2775 NOK
DALL6551131/42
Safety ankle boots for ladies, lace-up, Temptation® City Top
Elegant ladies ankle boots made from full grain leather, featuring leather lining, a leather insole and beige laces. The shoes provide high resistance to fuels and oils.
  • Flexible, light PU sole provides slip resistance on different types of surfaces
  • Protective steel toecap (200 J)
  • Metal-free, light and flexible puncture resistant Flexium™ midsole
For use in chemical industries, services, and logistics and transport.
Colour: Black
Sertifiseringer: EN ISO 20345:2007; S3, SRC.