Ultrasonic baths, Clifton
Leverandør: Nickel-Electro
NICKDU-8EA 29600 NOK
NICKDU-8 NICKMU-8 NICKDU-22 NICKDU4 NICKDU-14
Ultrasonic baths, Clifton
These ultrasonic baths are suitable for sonochemistry, degassing and sample dissolution and cleaning of delicate or fragile components/glassware especially where there may be a risk of breakage as a result of manual cleaning.
  • Totally unobstructed one piece, crevice free, easy to clean stainless steel tank
  • Bench top footprint
  • Comprising of a high quality corrosion resistant stainless steel tank
  • Sturdy construction - powder coated exterior providing a durable chemical resistant
  • IP rated clear booted on/off mains switch and bath fitted with non-slip rubber feet
  • Built to latest safety standards and supplied with comprehensive operating instructions
  • Each bath is uniquely identified with its own serial number recorded on our product database, which includes all of its manufacturing inspection data
  • Electrically tested for safety at final inspection and a test certificate enclosed in rear of the instruction book
Using high performance piezoelectric disc transducers to ensure consistent ultrasonic activity throughout the tank liquid ideal for laboratory routines. The activity is called cavitation created by transducers changing pressure waves causing bubbles to form and collapse.
It features mains on/off switch, timer and push button, illuminates and activates timer.
Heaters, transducers and temperature sensor concealed under the tank providing a clutter free working area easy to wipe clean. Wipe clean controls feature bold LED display of actual temperature, adjusting temperature and time and activating sonics. When a low liquid level is detected a “FiLL” message automatically appears, heating and ultrasonics are suspended. Over and under temperature alarms automatically protect work in the water bath, providing heater cut off, visual message and indicator.