Disperser system, ULTRA-TURRAX® Tube Drive control (UTTD control)
Leverandør: IKA
412-0161EA 0 NOK
IKAA3827500
Disperser system, ULTRA-TURRAX® Tube Drive control (UTTD control)
Universal dispersing, stirring, homogenising and grinding system with hermetically sealable disposable sample tubes. The UTTD control provides protection and security for infectious sample materials, toxic substances and high odour substances all under defined conditions of time, energy and volume.
  • Digital display for all functions including actual speed and target speed
  • USB interface for control and documentation
  • Adjustable reverse operation and Turbo button for short periods of intensive mixing, homogenising or grinding
  • A range of disposable sample tubes 20 and 50 ml including versions that are gamma sterilised and also tubes with piercable membrane closures
  • Hygienic and clean operation without any possible cross-contamination
  • Collecting tray for protection against any possible leaking liquids
Ideal for use in medical, pathology, toxicology, pharmaceutical, immunology, veterinary, clinical, food and cosmetics applications. The sample containers (tubes) are easily attached to the drive unit with a simple twist-lock coupling. Simple and precise multilingual menu navigation thanks to the OLED display that prompts the user to set the desired speed and duration. An acoustic signal indicates completion of experiment. The tests are programmable (from a library of saved sample conditions), reproducible and no cross-contamination between the samples.
Sertifiseringer: IP protection class according to DIN EN 60529: IP 20
Leveringsinformasjon: ULTRA-TURRAX® Tube Drive is supplied with EU/UK/CH power supply, removable hook for removing the rotor stator unit, but without tubes, please order required accessories separately.
Workstation includes 1x ULTRA-TURRAX® Tube Drive control, disposable tube: ST-20, DT-20, and 2×BMT-20G/S and hook for removing the rotor stator unit.