Norway
Order Entry
Rotors for Sorvall™ LYNX centrifuges
Rotors for Sorvall™ LYNX centrifuges
Certificates
undefined
Rotors for Sorvall™ LYNX centrifuges
Product Information
  • Catalog #
    Supplier #
    Pk
    Availability
    Price
  • Fixed-angle rotor Fiberlite™ F10-4x1000 LEX, carbon fibre, 4×1000 ml, 10500⁻¹, 20584×g
    Fixed-angle rotor Fiberlite™ F10-4x1000 LEX, carbon fibre, 4×1000 ml, 10500⁻¹, 20584×g
    521-2670
    096-041075
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 096-041075
    Local Article Number: SORV096-041075
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Fixed-angle rotor Fiberlite™ F9-6x1000 LEX, carbon fibre, 6×1000 ml, 9000⁻¹, 17568×g (only for use with Sorvall LYNX 6000 centrifuge)
    Fixed-angle rotor Fiberlite™ F9-6x1000 LEX, carbon fibre, 6×1000 ml, 9000⁻¹, 17568×g (only for use with Sorvall LYNX 6000 centrifuge)
    521-1964
    096-061075
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 096-061075
    Local Article Number: SORV096-061075
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Fixed-angle rotor Fiberlite™ F14-6x250y, carbon fibre, 6×250 ml (62×135 mm), 23° angle, 30240×g
    Fixed-angle rotor Fiberlite™ F14-6x250y, carbon fibre, 6×250 ml (62×135 mm), 23° angle, 30240×g
    521-1965
    096-062075
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 096-062075
    Local Article Number: SORV096-062075
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Fixed-angle rotor Fiberlite™ F12-6x500 LEX, carbon fibre, 6×500 ml (70×160 mm), 20° angle, 24515×g
    Fixed-angle rotor Fiberlite™ F12-6x500 LEX, carbon fibre, 6×500 ml (70×160 mm), 20° angle, 24515×g
    521-1966
    096-062185
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 096-062185
    Local Article Number: SORV096-062185
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Fixed-angle rotor Fiberlite™ F12-6x500 LEX, carbon fibre, 6×500 ml (70×160 mm), 20° angle, 24471×g
    Fixed-angle rotor Fiberlite™ F12-6x500 LEX, carbon fibre, 6×500 ml (70×160 mm), 20° angle, 24471×g
    521-1967
    096-062375
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 096-062375
    Local Article Number: SORV096-062375
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Fixed angle rotor Fiberlite™ F21-8x50y, carbon fibre, with auto-lock, 8×50 ml, 20000 min⁻¹, 47850×g
    Fixed angle rotor Fiberlite™ F21-8x50y, carbon fibre, with auto-lock, 8×50 ml, 20000 min⁻¹, 47850×g
    SORV096-084275
    096-084275
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 096-084275
    Local Article Number: SORV096-084275
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Fixed angle rotor Fiberlite™ F20-12x50 LEX, carbon fibre, 12×50 ml, 25° angle, 20000 min⁻¹, 51428×g
    Fixed angle rotor Fiberlite™ F20-12x50 LEX, carbon fibre, 12×50 ml, 25° angle, 20000 min⁻¹, 51428×g
    SORV096-124375
    096-124375
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 096-124375
    Local Article Number: SORV096-124375
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Swinging bucket rotor BIOFlex HC Rotor, stainless steel, 4×1000 ml (126×1140 mm), horizontal, 7068×g
    Swinging bucket rotor BIOFlex HC Rotor, stainless steel, 4×1000 ml (126×1140 mm), horizontal, 7068×g
    521-1977
    75003000
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 75003000
    Local Article Number: SORV75003000
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Swinging bucket rotor BIOFlex HS Rotor, stainless steel, 4×400 ml, horizontal, 7000 min⁻¹, 10025×g
    Swinging bucket rotor BIOFlex HS Rotor, stainless steel, 4×400 ml, horizontal, 7000 min⁻¹, 10025×g
    SORV75003002
    75003002
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 75003002
    Local Article Number: SORV75003002
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Fixed angle rotor, A27-6x50, aluminium, 6×50 ml, angle 34°, 27000 min⁻¹, 79057×g
    Fixed angle rotor, A27-6x50, aluminium, 6×50 ml, angle 34°, 27000 min⁻¹, 79057×g
    SORV75003007
    75003007
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 75003007
    Local Article Number: SORV75003007
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Fixed angle rotor, A27-8x50, aluminium, 8×50 ml, angle 34°, 27000 min⁻¹, 87207×g
    Fixed angle rotor, A27-8x50, aluminium, 8×50 ml, angle 34°, 27000 min⁻¹, 87207×g
    SORV75003008
    75003008
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 75003008
    Local Article Number: SORV75003008
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Swinging bucket rotor TH13-6x50, aluminum with titanium buckets, angle 90°, 30314×g
    Swinging bucket rotor TH13-6x50, aluminum with titanium buckets, angle 90°, 30314×g
    SORV75003010
    75003010
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 75003010
    Local Article Number: SORV75003010
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Fixed angle rotors, Fiberlite™ F14-6x250y, carbon fibre, 6x250 ml, 14000⁻¹, 30240×g
    Fixed angle rotors, Fiberlite™ F14-6x250y, carbon fibre, 6x250 ml, 14000⁻¹, 30240×g
    SORVAVL096-062075
    AVL096-062075
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: AVL096-062075
    Local Article Number: SORVAVL096-062075
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Fixed-angle rotor Fiberlite™ F14-14x50cy, carbon fibre, 14×50 ml (29×115 mm), 34° angle, 33746×g
    Fixed-angle rotor Fiberlite™ F14-14x50cy, carbon fibre, 14×50 ml (29×115 mm), 34° angle, 33746×g
    521-1968
    AVL096-145075
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: AVL096-145075
    Local Article Number: SORVAVL096-145075
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable