Cation exchange bulk packing material
Leverandør: Hamilton
Polymer bulk packing material for cation exchange applications.
Chromatographic capacity and efficiency tests are conducted on both bulk and packed column resins to ensure product integrity.
BESTILL
Utvid alle / Skjul alle
Beskrivelse Partikkelstr Porestr Fp Art.nr Antall
PRP-X200 bulk resin 10 µm 100 Å 1 g 553-0008
PRP-X200 bulk resin 12 - 20 µm 100 Å 1 g 553-0009
PRP-X400 bulk resin 7 µm - 1 g 553-0012
PRP-X400 bulk resin 12 - 20 µm - 1 g 553-0013
PRP-X400 bulk resin 30 - 50 µm - 1 g 553-0014
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall