Capillary columns, TRACE™ TR-1
Leverandør: Thermo Scientific
TRACE™
TRACE™ TR-1 is a general purpose 100% dimethylpolysiloxane non-polar column suitable for a wide range of compounds, including hydrocarbons, esters, pesticides, drugs and amines.
  • Low bleed for increased sensitivity
  • High operating temperature for easy reconditioning
  • Solvent washable
Logg inn for å se avtalepris og tilgjengelighet
  Husk meg på denne enheten
BESTILL
Utvid alle / Skjul alle
Column format Phase Tykkhet film Innvendig-Ø×L Fp Art.nr Antall
Capillary TR-1 0,10 µm 0,25 mm × 30 m 1 555-9352
Capillary TR-1 0,25 µm 0,25 mm × 15 m 1 555-9350
Capillary TR-1 0,25 µm 0,32 mm × 15 m 1 555-9285
Capillary TR-1 0,25 µm 0,25 mm × 30 m 1 555-9284
Capillary TR-1 0,25 µm 0,32 mm × 30 m 1 555-9283
Capillary TR-1 0,25 µm 0,25 mm × 60 m 1 555-9282
Capillary TR-1 0,50 µm 0,25 mm × 30 m 1 555-9277
Capillary TR-1 0,50 µm 0,25 mm × 100 m 1 555-9273
Capillary TR-1 1,00 µm 0,25 mm × 30 m 1 555-9272
Capillary TR-1 1,00 µm 0,32 mm × 30 m 1 555-9271
Capillary TR-1 1,00 µm 0,53 mm × 30 m 1 555-9270
Capillary TR-1 1,00 µm 0,25 mm × 60 m 1 555-9269
Capillary TR-1 1,50 µm 0,53 mm × 30 m 1 555-9266
Capillary TR-1 3,00 µm 0,32 mm × 30 m 1 555-9265
Capillary TR-1 3,00 µm 0,53 mm × 30 m 1 555-9264
Capillary TR-1 5,00 µm 0,32 mm × 30 m 1 555-9263
Capillary TR-1 5,00 µm 0,53 mm × 30 m 1 555-9262
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Velg produkt:
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall