Bottles, wide neck, round, with closure, Precisionware®
Leverandør: Bel-Art
Precisionware™
HDPE translucent. Flexible bottles provide a leakproof and easy filling and pouring design.
  • Good chemical resistance
  • Good temperature resistance
Supplied with matched polypropylene closures, these bottles offer greater visibility than HDPE bottles. Constructed from FDA compliant material, their wide mouth design makes general storage of liquids or powders simple. Bottles 250 mL and larger feature a recycling code molded into the base.
BESTILL
Utvid alle / Skjul alle
Kapasitet Gjenge Ytre diam Fp Materiale Art.nr Antall
1000 ml 53 mm 90,42 mm 6 LDPE BELAF106260018
125 ml 38 mm 48,26 mm 12 LDPE BELAF106260005
250 ml 45 mm 58,42 mm 12 LDPE BELAF106260006
500 ml 53 mm 72,14 mm 12 LDPE BELAF106260007
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall