DNA extraction kits, ZyGEM™ prepGEM® Saliva kit
ZYGEPSA0050EA 1550 NOK
ZYGEPSA0050
DNA extraction kits, ZyGEM™ prepGEM® Saliva kit
prepGEM® DNA extraction kits have been developed to provide a rapid, reliable, and economical solution to DNA extraction. The kits use a single tube protocol with temperature control to prepare DNA for analysis, and are optimised for downstream PCR-based applications, including qPCR, STR-PCR, DNA sequencing, AFLP analysis, and many SNiP detection applications.
prepGEM® Saliva kit is a single tube, enzymatic DNA extraction system optimised to extract DNA from human buccal swab samples, or liquid saliva samples, collected under laboratory, biomedical, or remote settings. The kits yield PCR ready DNA in less than 25 minutes, sufficient for multiple assays. The extracted DNA is suitable for PCR applications including qPCR, SNP detection, and routine genotyping, or whenever rapid results are required.