Heat resistant welding gloves, Argon
Leverandør: Honeywell Safety
112-1445EA 240 NOK
112-1445 112-1449 112-1446
Heat resistant welding gloves, Argon
Hansker
Goat leather grain glove with long split leather cuff (150 mm) and sewn-on thumb.
  • Assembly and flexible materials make these gloves anatomical, thus providing excellent comfort
  • Split leather provides excellent resistance to fire and convective heat
  • Leather cuff protects a large part of the forearm against mechanical hazards
Ideal for argon welding and fine handling.
Sertifiseringer: EN 388:2016 (3121X); Cat. II.