asecos, safety storage cabinets, S-Classic
Supplier: asecos
Certificates
About this item
Fireproof safety cabinets, classified type 90 (fire resistant up to 90 minutes).
- Robust construction and longevity - safety elements assembled outside the storage compartment for increased protection against corrosion, easy to clean
- Convenient handling: Doors can be opened with minimal effort
- User friendly and safe: Door open arrest system (optional), convenient and safe removal or storage of containers
- No unauthorised use: Doors lockable with cylinder locking (integration in a locking system possible)
- Easy alignment: Adjusting aids to compensate for uneven floor
- Quickly put into operation: Integrated air ducts ready for connection (NW 75 mm) to a technical exhaust system
Outer body made of sheet steel with high grade structured paint finish, interior with chemically resistant surface.
Certifications: Furnace tested (type test) in accordance with EN 14470-1
Fire resistance 90 minutes
GS approval, CE conformity
EN 14727 conformity (laboratory furniture)
Made in Germany
Fire resistance 90 minutes
GS approval, CE conformity
EN 14727 conformity (laboratory furniture)
Made in Germany
Ordering information: Other colours are available on request. Supplied without interior equipment packages. Interior equipment packages need to be ordered separately. Please check the website or contact Avantor for further information.
Delivery: Delivery prices available upon request.
Product Information
- Catalog #Supplier #DoorsW×D×HW×D×H int.WeightPkAvailabilityPrice
- Safety storage cabinet, type 90, without interior equipment, model S90.129.060, S-CLASSIC-90
Body and doors: light grey RAL 7035Safety storage cabinet, type 90, without interior equipment, model S90.129.060, S-CLASSIC-90
Body and doors: light grey RAL 7035135-2576302380011 wing (left hinged)596×616×1298 mm450×524×1072 mm170 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, model S90.129.060, S-CLASSIC-90
Body and doors: warning yellow RAL 1004Safety storage cabinet, type 90, without interior equipment, model S90.129.060, S-CLASSIC-90
Body and doors: warning yellow RAL 1004135-2577302380021 wing (left hinged)596×616×1298 mm450×524×1072 mm170 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.129.060.WDAS, S-CLASSIC-90
Body and doors: light grey RAL 7035Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.129.060.WDAS, S-CLASSIC-90
Body and doors: light grey RAL 7035135-2578302390011 wing (left hinged)596×616×1298 mm450×524×1072 mm170 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.129.060.WDAS, S-CLASSIC-90
Body and doors: warning yellow RAL 1004Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.129.060.WDAS, S-CLASSIC-90
Body and doors: warning yellow RAL 1004135-2579302390021 wing (left hinged)596×616×1298 mm450×524×1072 mm170 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, model S90.129.060.R, S-CLASSIC-90
Body and doors: light grey RAL 7035Safety storage cabinet, type 90, without interior equipment, model S90.129.060.R, S-CLASSIC-90
Body and doors: light grey RAL 7035135-2580302400011 wing (right hinged)596×616×1298 mm450×524×1072 mm170 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, model S90.129.060.R, S-CLASSIC-90
Body and doors: warning yellow RAL 1004Safety storage cabinet, type 90, without interior equipment, model S90.129.060.R, S-CLASSIC-90
Body and doors: warning yellow RAL 1004135-2581302400021 wing (right hinged)596×616×1298 mm450×524×1072 mm170 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.129.060.WDASR, S-CLASSIC-90
Body and doors: light grey RAL 7035Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.129.060.WDASR, S-CLASSIC-90
Body and doors: light grey RAL 7035135-2582302410011 wing (right hinged)596×616×1298 mm450×524×1072 mm170 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.129.060.WDASR, S-CLASSIC-90
Body and doors: warning yellow RAL 1004Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.129.060.WDASR, S-CLASSIC-90
Body and doors: warning yellow RAL 1004135-2583302410021 wing (right hinged)596×616×1298 mm450×524×1072 mm170 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, model S90.196.090, S-CLASSIC-90
Body and doors: light grey RAL 7035Safety storage cabinet, type 90, without interior equipment, model S90.196.090, S-CLASSIC-90
Body and doors: light grey RAL 7035135-1697302270011 wing (left hinged)596×616×1968 mm450×524×1742 mm260 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, model S90.196.060, S-CLASSIC-90
Body and doors: warning yellow RAL 1004Safety storage cabinet, type 90, without interior equipment, model S90.196.060, S-CLASSIC-90
Body and doors: warning yellow RAL 1004135-1695302270021 wing (left hinged)596×616×1968 mm450×524×1742 mm260 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.196.060.WDAS, S-CLASSIC-90
Body and doors: light grey RAL 7035Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.196.060.WDAS, S-CLASSIC-90
Body and doors: light grey RAL 7035135-2570302280011 wing (left hinged)596×616×1968 mm450×524×1742 mm260 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.196.060.WDAS, S-CLASSIC-90
Body and doors: warning yellow RAL 1004Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.196.060.WDAS, S-CLASSIC-90
Body and doors: warning yellow RAL 1004135-2571302280021 wing (left hinged)596×616×1968 mm450×524×1742 mm260 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, model S90.196.060.R, S-CLASSIC-90
Body and doors: light grey RAL 7035Safety storage cabinet, type 90, without interior equipment, model S90.196.060.R, S-CLASSIC-90
Body and doors: light grey RAL 7035135-2572302290011 wing (right hinged)596×616×1968 mm450×524×1742 mm260 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, model S90.196.060.R, S-CLASSIC-90
Body and doors: warning yellow RAL 1004Safety storage cabinet, type 90, without interior equipment, model S90.196.060.R, S-CLASSIC-90
Body and doors: warning yellow RAL 1004135-2573302290021 wing (right hinged)596×616×1968 mm450×524×1742 mm260 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.196.060.WDASR, S-CLASSIC-90
Body and doors: light grey RAL 7035Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.196.060.WDASR, S-CLASSIC-90
Body and doors: light grey RAL 7035135-2574302300011 wing (right hinged)596×616×1968 mm450×524×1742 mm260 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.196.060.WDASR, S-CLASSIC-90
Body and doors: warning yellow RAL 1004Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.196.060.WDASR, S-CLASSIC-90
Body and doors: warning yellow RAL 1004135-2575302300021 wing (right hinged)596×616×1968 mm450×524×1742 mm260 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, model S90.129.120, S-CLASSIC-90
Body and doors: light grey RAL 7035Safety storage cabinet, type 90, without interior equipment, model S90.129.120, S-CLASSIC-90
Body and doors: light grey RAL 7035135-2067301210012 wing1196×616×1298 mm1050×524×1072 mm270 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, model S90.196.120, S-CLASSIC-90
Body and doors: warning yellow RAL 1004Safety storage cabinet, type 90, without interior equipment, model S90.196.120, S-CLASSIC-90
Body and doors: warning yellow RAL 1004135-2068301210022 wing1196×616×1298 mm1050×524×1072 mm270 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.129.120.WDAS, S-CLASSIC-90
Body and doors: light grey RAL 7035Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.129.120.WDAS, S-CLASSIC-90
Body and doors: light grey RAL 7035135-2566301220012 wing1196×616×1298 mm1050×524×1072 mm270 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.129.120.WDAS, S-CLASSIC-90
Body and doors: warning yellow RAL 1004Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.129.120.WDAS, S-CLASSIC-90
Body and doors: warning yellow RAL 1004135-2567301220022 wing1196×616×1298 mm1050×524×1072 mm270 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, Model S-90.196.090, S-CLASSIC
Body and doors: light grey RAL 7035Safety storage cabinet, type 90, without interior equipment, Model S-90.196.090, S-CLASSIC
Body and doors: light grey RAL 7035135-1761302040012 wing896×616×1968 mm750×524×1742 mm310 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, model S90.196.090, S-CLASSIC-90
Body and doors: warning yellow RAL 1004Safety storage cabinet, type 90, without interior equipment, model S90.196.090, S-CLASSIC-90
Body and doors: warning yellow RAL 1004135-1762302040022 wing896×616×1968 mm750×524×1742 mm310 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.196.090.WDAS, S-CLASSIC-90
Body and doors: light grey RAL 7035Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.196.090.WDAS, S-CLASSIC-90
Body and doors: light grey RAL 7035135-2568302050012 wing896×616×1968 mm750×524×1742 mm310 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.196.090.WDAS, S-CLASSIC-90
Body and doors: warning yellow RAL 1004Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.196.090.WDAS, S-CLASSIC-90
Body and doors: warning yellow RAL 1004135-2569302050022 wing896×616×1968 mm750×524×1742 mm310 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.196.060.075.WDAS, S-CLASSIC-90
Body and doors: pure white RAL 9010Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.196.060.075.WDAS, S-CLASSIC-90
Body and doors: pure white RAL 9010135-2585325320051 wing596×749×1968 mm450×657×1742 mm310 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.196.060.075.WDASR, S-CLASSIC-90
Body and doors: pure white RAL 9010Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.196.060.075.WDASR, S-CLASSIC-90
Body and doors: pure white RAL 9010135-2586325330051 wing596×749×1968 mm450×657×1742 mm310 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, model S90.196.120, S-CLASSIC-90
Body and doors: light grey RAL 7035Safety storage cabinet, type 90, without interior equipment, model S90.196.120, S-CLASSIC-90
Body and doors: light grey RAL 7035135-1759301150012 wing1196×616×1968 mm1050×524×1742 mm420 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, model S90.196.120, S-CLASSIC-90
Body and doors: warning yellow RAL 1004Safety storage cabinet, type 90, without interior equipment, model S90.196.120, S-CLASSIC-90
Body and doors: warning yellow RAL 1004135-1760301150022 wing1196×616×1968 mm1050×524×1742 mm420 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, model S90.196.120, S-CLASSIC-90
Body and doors: warning light grey RAL 7035Safety storage cabinet, type 90, without interior equipment, model S90.196.120, S-CLASSIC-90
Body and doors: warning light grey RAL 7035135-2564301160012 wing1196×616×1968 mm1050×524×1742 mm420 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable - Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.196.120.WDAS, S-CLASSIC-90
Body and doors: warning yellow RAL 1004Safety storage cabinet, type 90, without interior equipment, with door open arrest, model S90.196.120.WDAS, S-CLASSIC-90
Body and doors: warning yellow RAL 1004135-2565301160022 wing1196×616×1968 mm1050×524×1742 mm420 kg1Log in for availability1 * 1 ItemsSpecifications:Accessories
Specifications
Environmentally Preferable