3-Amino-6-bromo-1H-indazole 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:404827-77-6
:
Formula:C₇H₆BrN₃
MDL Number:MFCD05665872
:212.05 g/mol
Melting Pt:227…230 °C
:GHS07
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: H34095.03Local Article Number: ALFAH34095.03
- Supplier Article Number: H34095.09Local Article Number: ALFAH34095.09