3-Aminobenzoic acid hydrochloride 98%
|
Warning
|
L08407.22EA
1075
NOK
L08407.22
3-Aminobenzoic acid hydrochloride 98%
3-Aminobenzoic acid hydrochloride
Formel:
C₇H₈ClNO₂ MW: 173,60 g/mol Smeltepunkt: ∼ 286 °C (dec.) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00012973 CAS-nummer: 15151-51-6 EINECS: 239-212-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...