Norway
Order Entry
Blood lancets, VITREX® STERIHEEL® Baby II
undefined
Blood lancets, VITREX® STERIHEEL® Baby II
  •  
    Colour code
    Application
    Size
    Pk
  • MODU335213
    335213
    Green
    Micro-Preemie
    0,65x1,40 mm
    50
    1 * 50 Items
    1 * 50 Items - kr 1 900,00
    kr 1 900,00
    1 * 50 Items
       kr 1 900,00
    Specifications
    Environmentally Preferable
  • MODU334213
    334213
    Orange
    Preemie
    0,85x1,75 mm
    50
    1 * 50 Items
    1 * 50 Items - kr 1 900,00
    kr 1 900,00
    1 * 50 Items
       kr 1 900,00
    Specifications
    Environmentally Preferable
  • MODU333213
    333213
    Yellow
    Newborn
    1,00x2,50 mm
    50
    1 * 50 Items
    1 * 50 Items - kr 1 900,00
    kr 1 900,00
    1 * 50 Items
       kr 1 900,00
    Specifications
    Environmentally Preferable
  • MODU332213
    332213
    Pink
    Newborn Plus
    1,14x2,80 mm
    50
    1 * 50 Items
    1 * 50 Items - kr 1 900,00
    kr 1 900,00
    1 * 50 Items
       kr 1 900,00
    Specifications
    Environmentally Preferable
  • MODU331213
    331213
    Blue
    Toddler
    2,00x3,00 mm
    50
    1 * 50 Items
    1 * 50 Items - kr 1 900,00
    kr 1 900,00
    1 * 50 Items
       kr 1 900,00
    Specifications
    Environmentally Preferable