Heating cabinet for eyewash solutions
Leverandør: Orkla
CEDE790400EA 6300 NOK
CEDE790400
Heating cabinet for eyewash solutions
Førstehjelpsutstyr Øyeskyllestasjon
This cabinet is intended for the storage of two 500 ml bottles of Cederroth eye wash in cold environments, keeping the temperature of the eye wash solution at +20 °C (±2°C) when the surrounding temperature is between −20 °C and +20 °C.
  • Made of isolating expanded polypropylene (EPP) with a thermostatic heating element
  • Cabinet automatically switches between 12V DC and 24V DC (at 230V a transformer is needed)
  • Cabinet is impact resistant, resistant against chemicals and temperatures up to 130 °C
  • Supplied with electrical cord (2 m) with plug for 12V/24V cigarette lighter outlets
W×D×H: 276×130×315 mm