Gå til Overskrift
Gå til Hovedinnhold
Gå til Bunntekst
Avansert
Avansert
SDS
Sertifikat
Produsentdata
Logg inn
/
Register
Norway
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
kr 0,00
Order Entry
Home
Trimellitic acid tripropyl ester
Trimellitic acid tripropyl ester ≥97.0% (by GC)
Trimellitic acid tripropyl ester ≥97.0% (by GC)
Trimellitic acid tripropyl ester ≥97.0% (by GC)
TCIAT1446-25G
:
TCI
:
Trimellitic acid tripropyl ester ≥97.0% (by GC)
TCIAT1446-25G
:
TCI
:
kr 1 275,00
1 * 25 g
-
+
Pack type:
Glass bottle
Pk:
25 g
Seq:
3308
Appearance:
Colorless to Light yellow clear liquid
Purity (GC):
min. 97.0 %
:
:
168°C
:
1528-53-6
:
C₁₈H₂₄O₆
:
MFCD00191686