Very high pressure PK fittings, Upchurch Scientific®
Ultra high performance fittings are manufactured from a proprietary PEEK blend (PK) which allow them to be used at higher temperatures (up to 200 °C) and higher pressures.
VHP PK one piece fittings
- Pk
- PK-120BLK, PK one-piece fitting, for 1/16" OD tubing, 10-32 coned port, 12000 psi, standard knurlPK-120BLK, PK one-piece fitting, for 1/16" OD tubing, 10-32 coned port, 12000 psi, standard knurlUPCHPK-120BLKPK-120BLK11 * 1 Itemskr 350,00: PK-120BLK: UPCHPK-120BLK
Specifications
Environmentally Preferable - PK-120BLKx, PK one-piece fitting, for 1/16" OD tubing, 10-32 coned port, 12000 psi, standard knurlPK-120BLKx, PK one-piece fitting, for 1/16" OD tubing, 10-32 coned port, 12000 psi, standard knurlUPCHPK-120BLKXPK-120BLKX101 * 10 Itemskr 3 325,00: PK-120BLKX: UPCHPK-120BLKX
Specifications
Environmentally Preferable - PK-126, PK one-piece fitting, for 1/32" OD tubing, 6-32 coned port, 15000 psi, standard micro knurlPK-126, PK one-piece fitting, for 1/32" OD tubing, 6-32 coned port, 15000 psi, standard micro knurlUPCHPK-126PK-12611 * 1 Itemskr 500,00: PK-126: UPCHPK-126
Specifications
Environmentally Preferable
VHP PK LiteTouch® fittings
- Pk
- PK-100x, PK ferrule, for 1/16" OD tubing, 10-32 coned port, 16500 psiPK-100x, PK ferrule, for 1/16" OD tubing, 10-32 coned port, 16500 psiUPCHPK-100XPK-100X101 * 10 Itemskr 3 175,00: PK-100X: UPCHPK-100X
Specifications
Environmentally Preferable - PK-110x, PK nut, for 1/16" OD tubing, 10-32 coned port, 16500 psi, standard knurlPK-110x, PK nut, for 1/16" OD tubing, 10-32 coned port, 16500 psi, standard knurlUPCHPK-110XPK-110X101 * 10 Itemskr 1 675,00: PK-110X: UPCHPK-110X
Specifications
Environmentally Preferable - PK-132, PK ferrule, for 1/32" OD tubing, 10-32 coned port, 16500 psiPK-132, PK ferrule, for 1/32" OD tubing, 10-32 coned port, 16500 psiUPCHPK-132PK-13211 * 1 Itemskr 485,00: PK-132: UPCHPK-132
Specifications
Environmentally Preferable