Hansker, for generell bruk, ActivArmr® Heavy Duty 97-009
Leverandør: Ansell
ActivArmr®
ANSE97-009/8EA 9250 NOK
ANSE97-009/8 ANSE97-009/11 ANSE97-009/9 ANSE97-009/10
Hansker, for generell bruk, ActivArmr® Heavy Duty 97-009
This rugged, versatile glove is engineered for the most demanding jobs. The knitted liner is made of stainless steel, Lycra® and Kevlar® fabric and features a nitrile foam coating on the palm.
  • Intercept Technology®, reinforced with stainless steel and Dupont™ Kevlar®, delivers ultimate protection from punctures, cuts and abrasions
  • Reinforced ridges offer additional palm and knuckle protection
  • Foam nitrile coating provides enhanced wet or dry grip
  • “Second skin” feel gives dexterity and enables precision work all day long
  • With adjustable wrist closure with pull-tab and ID nameplate
Ideal for general construction applications, handling sheet metal, demolition and wood work, heavy cutting/sawing and steel work (tying).
Sertifiseringer: EN 388 (4432), Cat. II