Norway
Order Entry
96-Well assay plates, 'V' well, Axygen®
96-Well assay plates, 'V' well, Axygen®
 :  Corning
undefined
96-Well assay plates, 'V' well, Axygen®
 :  Corning
  • Catalog No:
  • AXYGP-96-450V-B
  • AXYGP-96-450V-Y
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  •  
    Colour
    Working volume
    Total volume
    Pk
  • 96-well assay plate, 'V' well bottom
    96-well assay plate, 'V' well bottom
    AXYGP-96-450V-B
    P-96-450V-B
    Blue
    480 µl
    520 µl
    10
    1 * 10 Items
    1 * 10 Items - kr 355,00
    kr 355,00
    1 * 10 Items
       kr 355,00
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact
  • 96-well assay plate, 'V' well bottom
    96-well assay plate, 'V' well bottom
    AXYGP-96-450V-Y
    P-96-450V-Y
    Yellow
    480 µl
    520 µl
    10
    1 * 10 Items
    1 * 10 Items - kr 355,00
    kr 355,00
    1 * 10 Items
       kr 355,00
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact