Round cover glasses, Nunc Thermanox
Supplier: Thermo Fisher Scientific
Certificates
About this item
These cover glasses remain flat during normal use.
- Resistant to common solvents
- Autofluorescent from 380 to 545 nm
- Surfaced treatment on one side for optimal cell attachment and growth
Specifications
- Catalog No:
- 734-2117
- 734-2173
- 734-2118
- 734-2172
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Specifications
Product Information
- Catalog #Supplier #ThicknessØPkAvailabilityPrice
- Round coverslips for 24-well multidishRound coverslips for 24-well multidish734-21171749500,2 mm13 mm5001 * 500 Itemskr 6 675,00Supplier Article Number: 174950Local Article Number: NUNC174950Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - Round coverslipsRound coverslips734-21731749770,2 mm22 mm5001 * 500 Itemskr 10 950,00Supplier Article Number: 174977Local Article Number: NUNC174977Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - Round coverslips for 6-well multidishRound coverslips for 6-well multidish734-21181749850,2 mm25 mm5001 * 500 Itemskr 10 850,00Supplier Article Number: 174985Local Article Number: NUNC174985Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - Round coverslipsRound coverslips734-21721749690,2 mm15 mm5001 * 500 Itemskr 7 675,00Supplier Article Number: 174969Local Article Number: NUNC174969Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact